Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000450953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
ENST00000450953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039679.hg.1 |
TC01002551.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039679.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039679.hg.1 |
ENST00000429311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039679.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203319.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203319.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203319.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203319.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203319.hg.1 |
TC0100013811.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000372816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000397332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000450953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
ENST00000450953.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033081.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_001033082.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013811.hg.1 |
NM_005376.4 |
Refseq |