Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000486269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000486269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000486269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
ENST00000486269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007699.hg.1 |
ENST00000453566.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009240.hg.1 |
ENST00000423401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009240.hg.1 |
ENST00000423401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009240.hg.1 |
ENST00000423401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009240.hg.1 |
ENST00000423401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009240.hg.1 |
ENST00000423401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007013.hg.1 |
ENST00000297258.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007013.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007013.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007013.hg.1 |
TC08000518.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300009240.hg.1 |
ENST00000423401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009240.hg.1 |
ENST00000423401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009240.hg.1 |
ENST00000423401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009240.hg.1 |
ENST00000423401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009240.hg.1 |
ENST00000423401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155506.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155506.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155506.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155506.hg.1 |
ENST00000486269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155506.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800155506.hg.1 |
TC0800008080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800155506.hg.1 |
ENST00000486269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155506.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155506.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155506.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155506.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000297258.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000396359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000481695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000486269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000486269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000486269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
ENST00000486269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008080.hg.1 |
NM_001444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007699.hg.1 |
ENST00000453566.2 |
Ensembl |