Assay Details
Gene Symbol: CYP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000761.4 | NP_000752.2 | 3 | 949 | 68 | |
GenBank mRNA | AF182274.1 | - | 2 | 893 | 68 | |
BC067424.1 | - | 3 | 929 | 7002123 | 68 | |
BC067425.1 | - | 3 | 929 | 7002124 | 68 | |
BC067426.1 | - | 3 | 929 | 7002125 | 68 | |
BC067427.1 | - | 3 | 929 | 7002126 | 68 | |
BC067428.1 | - | 3 | 929 | 7002128 | 68 | |
BC067429.1 | - | 3 | 929 | 7002129 | 68 | |
M55053.1 | - | 2 | 893 | 68 | ||
Z00036.1 | - | 2 | 893 | 68 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
