Assay Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004720.2 | NP_001004720.1 | 3 | 857 | 71 | |
NM_003581.4 | NP_003572.2 | 4 | 967 | 71 | ||
XM_006712797.3 | XP_006712860.1 | 4 | 1623 | 71 | ||
XM_011511991.2 | XP_011510293.1 | 4 | 1061 | 71 | ||
XM_017005103.1 | XP_016860592.1 | 5 | 1225 | 71 | ||
XM_017005104.1 | XP_016860593.1 | 4 | 1237 | 71 | ||
XM_017005105.1 | XP_016860594.1 | 4 | 974 | 71 | ||
GenBank mRNA | AF043119.1 | - | 2 | 719 | 71 | |
AF047487.1 | - | 3 | 823 | 71 | ||
BC000103.2 | - | 3 | 741 | 3505427 | 71 | |
BC007195.1 | - | 3 | 748 | 3608149 | 71 | |
KU178418.1 | - | 2 | 691 | 71 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
