Assay Details
Gene Symbol: BCL2L15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY265863.1 | - | 1 - 2 | 126 | 76 | |
CR749373.1 | - | 1 - 2 | 213 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393320.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393320.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393320.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393320.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000393320.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000471267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000471267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000471267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000486485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000486485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000486485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000486485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000488450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000488450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000488450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | ENST00000488450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000369580.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393316.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393320.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393320.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393320.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393320.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000393320.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000471267.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000471267.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000471267.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000486485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000486485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000486485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000486485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000488450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000488450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000488450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | ENST00000488450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015336.hg.1 | NM_001010922.2 | Refseq |
