Assay Details
Gene Symbol: EDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005610.3 | NP_001005610.2 | 1 - 2 | 638 | 63 | |
GenBank mRNA | CA433299.1 | - | 1 - 2 | 63 | ||
U59228.1 | - | 1 - 2 | 638 | 63 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000365.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
