Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006387.hg.1 |
ENST00000225928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006387.hg.1 |
ENST00000225929.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006387.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006387.hg.1 |
TC17000528.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010728.hg.1 |
ENST00000590513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007896.hg.1 |
ENST00000590052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
TC1700007897.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157523.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000225929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000585807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000587280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000590299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
ENST00000593215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007897.hg.1 |
NM_000413.3 |
Refseq |