Assay Details
Gene Symbol: HIST1H2BA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170610.2 | NP_733759.1 | 1 | 30 | 104 | |
GenBank mRNA | AK312055.1 | - | 1 | 162 | 104 | |
BC066238.1 | - | 1 | 30 | 7002047 | 104 | |
BC066239.1 | - | 1 | 30 | 7002048 | 104 | |
BC066240.1 | - | 1 | 30 | 7002052 | 104 | |
BC066241.1 | - | 1 | 30 | 7002054 | 104 | |
BC066242.1 | - | 1 | 30 | 7002055 | 104 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000158.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
