Assay Details
Gene Symbol: MCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182763.2 | NP_877495.1 | 1 - 2 | 903 | 98 | |
GenBank mRNA | AF203373.1 | - | 1 - 2 | 695 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cell fate determination
multicellular organism development
intrinsic apoptotic signaling pathway in response to DNA damage
apoptotic mitochondrial changes
cellular homeostasis
response to cytokine
negative regulation of apoptotic process
protein transmembrane transport
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
negative regulation of anoikis
regulation of response to DNA damage stimulus
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of intrinsic apoptotic signaling pathway
protein binding
protein transmembrane transporter activity
protein homodimerization activity
protein heterodimerization activity
BH3 domain binding
cell fate determination
multicellular organism development
intrinsic apoptotic signaling pathway in response to DNA damage
apoptotic mitochondrial changes
cellular homeostasis
response to cytokine
negative regulation of apoptotic process
protein transmembrane transport
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
negative regulation of anoikis
regulation of response to DNA damage stimulus
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of intrinsic apoptotic signaling pathway
protein binding
protein transmembrane transporter activity
protein homodimerization activity
protein heterodimerization activity
BH3 domain binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243075.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243075.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243075.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243075.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243075.hg.1 | TC0100018484.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100243075.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243075.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243075.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243075.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | TC0100018484.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243076.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | TC0100018484.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243074.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243077.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243077.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243077.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243077.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243077.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243077.hg.1 | TC0100018484.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100243077.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243077.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100243077.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243077.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100243077.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000307940.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000369026.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000464132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | ENST00000620947.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_001197320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_021960.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018484.hg.1 | NM_182763.2 | Refseq |
