Assay Details
Gene Symbol: CMKLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142343.1 | NP_001135815.1 | 1 - 2 | 287 | 54 | |
NM_004072.2 | NP_004063.1 | 1 - 2 | 287 | 54 | ||
GenBank mRNA | BC106927.2 | - | 1 - 2 | 101 | 40033186 | 54 |
BC106928.1 | - | 1 - 2 | 101 | 40033187 | 54 | |
U79527.1 | - | 1 - 2 | 293 | 54 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001933.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC106927.2, BC106928.1, NM_001142343.1, NM_004072.2, U79527.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
chemotaxis
immune response
G protein-coupled receptor signaling pathway
positive regulation of macrophage chemotaxis
negative regulation of NF-kappaB transcription factor activity
negative regulation of interleukin-12 production
positive regulation of fat cell differentiation
regulation of calcium-mediated signaling
chemokine-mediated signaling pathway
G protein-coupled receptor activity
chemokine receptor activity
protein binding
signaling receptor activity
skeletal system development
chemotaxis
immune response
G protein-coupled receptor signaling pathway
positive regulation of macrophage chemotaxis
negative regulation of NF-kappaB transcription factor activity
negative regulation of interleukin-12 production
positive regulation of fat cell differentiation
regulation of calcium-mediated signaling
chemokine-mediated signaling pathway
G protein-coupled receptor activity
chemokine receptor activity
protein binding
signaling receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12025144.hg.1 | ENST00000550402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12025144.hg.1 | ENST00000552995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12025144.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12025144.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12025144.hg.1 | TC12001933.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191683.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191683.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191683.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191683.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191683.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191683.hg.1 | TC1200011838.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200191683.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191683.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191683.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
