Assay Details
Gene Symbol: SNORD114-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003223.1 | 1 | 36 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
small nucleolar RNA, C/D box 114-29 |
SNORD114-29 | 14q(II-29) | -1.8kb | Sense | Chr. 14 - 100990091 - 100990159 on NCBI Build GRCh38 |
Downstream |
small nucleolar RNA, C/D box 114-31 |
SNORD114-31 | 14q(II-31) | 1.2kb | Sense | Chr. 14 - 100993236 - 100993309 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000710.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4140 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29979867 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29979864 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09517534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29979862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09517538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29979866 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09517538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09517534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29979864 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29979862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29979866 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29979867 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517536 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517571 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517576 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517575 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517570 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517564 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517595 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16734936 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517480 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517486 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517585 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517554 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517573 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517489 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517459 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517533 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517528 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517568 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517606 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517520 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517578 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517540 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517559 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517589 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517493 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517602 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517535 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517517 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517465 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517601 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517583 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517588 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517557 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517587 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517482 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517593 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517484 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517609 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517538 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517597 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517581 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517543 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517599 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517498 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517514 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517604 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09517561 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400010688.hg.1 | ENST00000364448.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010688.hg.1 | NR_003223.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14008161.hg.1 | ENST00000364448.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14008161.hg.1 | NR_003223.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14008161.hg.1 | TC14000710.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1400180763.hg.1 | ENST00000364448.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400180763.hg.1 | NR_003223.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400180763.hg.1 | TC1400010688.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400180763.hg.1 | NR_003223.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400180763.hg.1 | ENST00000364448.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010688.hg.1 | ENST00000364448.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010688.hg.1 | NR_003223.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3310050 | NR_003223.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3310050 | NR_003223.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310050 | NR_003223.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310050 | NR_003223.1 | human_transcriptome |
