Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019064.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019064.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019064.hg.1 |
TC0X001440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019049.hg.1 |
ENST00000358993.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019049.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019049.hg.1 |
TC0X002342.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011402.hg.1 |
ENST00000370519.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011402.hg.1 |
ENST00000370519.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011402.hg.1 |
ENST00000370519.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011402.hg.1 |
ENST00000370519.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011402.hg.1 |
ENST00000370519.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011402.hg.1 |
NM_013453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011402.hg.1 |
NM_013453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011402.hg.1 |
NM_013453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011402.hg.1 |
NM_013453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011403.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011403.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011403.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011403.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011403.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011403.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011403.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011403.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011403.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011403.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008608.hg.1 |
ENST00000370518.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008608.hg.1 |
ENST00000370518.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008608.hg.1 |
ENST00000370518.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008608.hg.1 |
ENST00000370518.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008608.hg.1 |
ENST00000370518.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008608.hg.1 |
NM_145662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008608.hg.1 |
NM_145662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008608.hg.1 |
NM_145662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008608.hg.1 |
NM_145662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010990.hg.1 |
ENST00000358993.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010990.hg.1 |
ENST00000358993.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010990.hg.1 |
ENST00000358993.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010990.hg.1 |
ENST00000358993.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010990.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010990.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010990.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010990.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010990.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019063.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019063.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019063.hg.1 |
TC0X001440.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0X00011403.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011403.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011403.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011403.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011403.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011403.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011403.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011403.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011403.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011403.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010990.hg.1 |
ENST00000358993.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010990.hg.1 |
ENST00000358993.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010990.hg.1 |
ENST00000358993.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010990.hg.1 |
ENST00000358993.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010990.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010990.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010990.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010990.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010990.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174665.hg.1 |
ENST00000358993.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174665.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174665.hg.1 |
TC0X00010990.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00174665.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174665.hg.1 |
ENST00000358993.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174666.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174666.hg.1 |
TC0X00010990.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00174666.hg.1 |
NM_022661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00179145.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00179145.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00179145.hg.1 |
TC0X00011403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00179145.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00179145.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174667.hg.1 |
TC0X00010990.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00179147.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00179147.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00179147.hg.1 |
TC0X00011403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00179147.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00179147.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011402.hg.1 |
ENST00000370519.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011402.hg.1 |
ENST00000370519.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011402.hg.1 |
ENST00000370519.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011402.hg.1 |
ENST00000370519.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011402.hg.1 |
ENST00000370519.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011402.hg.1 |
NM_013453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011402.hg.1 |
NM_013453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011402.hg.1 |
NM_013453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011402.hg.1 |
NM_013453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008608.hg.1 |
ENST00000370518.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008608.hg.1 |
ENST00000370518.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008608.hg.1 |
ENST00000370518.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008608.hg.1 |
ENST00000370518.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008608.hg.1 |
ENST00000370518.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008608.hg.1 |
NM_145662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008608.hg.1 |
NM_145662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008608.hg.1 |
NM_145662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008608.hg.1 |
NM_145662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00179144.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00179144.hg.1 |
NM_032417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00179144.hg.1 |
TC0X00011403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00179144.hg.1 |
ENST00000370515.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00179144.hg.1 |
NM_032417.3 |
Refseq |