Assay Details
Gene Symbol: LINC00222 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY927449.1 | - | LIT3477 AY927449 |
4 - 5 | 534 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | forkhead box O3 | FOXO3 |
AF6q21, FKHRL1, FKHRL1P2, FOXO2, FOXO3A |
-82.2kb | Sense | Chr. 6 - 108559823 - 108684769 on NCBI Build GRCh38 |
Downstream | zinc finger protein 259 pseudogene 1 | ZNF259P1 |
354J5, ZNF259P |
16.9kb | Anti-sense | Chr. 6 - 108785327 - 108787059 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
