Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
1556730_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008235.hg.1 |
ENST00000547721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008235.hg.1 |
ENST00000547721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008235.hg.1 |
ENST00000547721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008235.hg.1 |
ENST00000547721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000266671.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000602540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
ENST00000619060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011310.hg.1 |
NM_007350.3 |
Refseq |
Human Genome U133 Plus 2.0 Array |
217998_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1556730_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
217998_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1556730_PM_at |
- |
- |
Human Genome U133A 2.0 Array |
217998_PM_at |
- |
- |
Human Genome U133A 2.0 Array |
217998_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556730_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556730_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556730_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556730_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556730_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556730_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556730_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556730_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556730_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556730_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556730_at8 |
- |
- |
Human Genome U133 Set |
217998_PM_at |
- |
- |
Human Genome U133 Set |
217998_at |
- |
- |