Assay Details
Gene Symbol: FBXW8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC045190.2 | - | 1 | 1946 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
harakiri, BCL2 interacting protein |
HRK |
DP5, HARAKIRI |
-67.3kb | Sense | Chr. 12 - 116856144 - 116881427 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC100506551 | LOC100506551 | 26.0kb | Sense | Chr. 12 - 116977440 - 116987337 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
