Assay Details
Gene Symbol: LINC01498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX648662.1 | - | 4 - 5 | 349 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream | FIC domain containing | FICD |
HIP13, HYPE, UNQ3041 |
29.0kb | Anti-sense | Chr. 12 - 108515274 - 108519603 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | NR_120483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | NR_120483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | NR_120483.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | NR_120483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | NR_120483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | NR_120483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | NR_120483.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | NR_120483.1 | Refseq |
