Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000433061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000433061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000433061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000433061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000433061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000433061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000433061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000433061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000433061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000433061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000608623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
ENST00000609401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011370.hg.1 |
NR_015353.1 |
Refseq |