Assay Details
Gene Symbol: FLJ42351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033871.1 | 2 - 3 | 138 | 73 | ||
GenBank mRNA | AK124342.1 | - | 2 - 3 | 1648 | 73 | |
AW027944.1 | - | 3 - 4 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil-helix-coiled-coil-helix domain containing 5 | CHCHD5 |
C2orf9, MIC14 |
-54.1kb | Anti-sense | Chr. 2 - 112583606 - 112589040 on NCBI Build GRCh38 |
Downstream | solute carrier family 20 member 1 | SLC20A1 |
GLVR1, Glvr-1, PIT1, PiT-1 |
1.8kb | Anti-sense | Chr. 2 - 112645857 - 112663825 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002216.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
