Assay Details
Gene Symbol: LOC100129675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK125061.1 | - | 1 | 53 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
BOK, BCL2 family apoptosis regulator |
BOK |
BCL2L9, BOKL |
-1.4kb | Sense | Chr. 2 - 241558731 - 241574138 on NCBI Build GRCh38 |
Downstream | THAP domain containing 4 | THAP4 |
CGI-36, PP238 |
5.2kb | Anti-sense | Chr. 2 - 241584405 - 241637543 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
