Assay Details
Gene Symbol: UNQ6494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK126399.1 | - | 1 | 2448 | 68 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | DB351680.1 | - | 1 | Chr. 9 - 89642170 - 89642394 [-] on Build GRCh38 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | growth arrest and DNA damage inducible gamma | GADD45G |
CR6, DDIT2, GADD45gamma, GRP17 |
-33.2kb | Sense | Chr. 9 - 89605009 - 89606555 on NCBI Build GRCh38 |
Downstream | putative uncharacterized protein UNQ6494/PRO21346 | LOC102724237 | 22.5kb | Anti-sense | Chr. 9 - 89661915 - 89672876 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
