Assay Details
Gene Symbol: WNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011541597.2 | XP_011539899.1 | 1 | 953 | 78 | |
GenBank mRNA | AF086347.1 | - | 1 | 453 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cell division cycle 42 | CDC42 |
CDC42Hs, G25K, TKS |
-48.3kb | Anti-sense | Chr. 1 - 22052627 - 22092943 on NCBI Build GRCh38 |
Downstream | microRNA 4418 | MIR4418 | 124.4kb | Anti-sense | Chr. 1 - 22266239 - 22266300 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_011541597.2 transcripts.
This assay is in the 5'UTR region of the XM_011541597.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
