Assay Details
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL360187.1 | - | 1 | 653 | 75 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AA680400.1 | - | 1 | Chr. 11 - 5696071 - 5696527 [+] on Build GRCh38 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tripartite motif containing 5 | TRIM5 |
RNF88, TRIM5alpha |
-10.9kb | Sense | Chr. 11 - 5590906 - 5685109 on NCBI Build GRCh38 |
Downstream | olfactory receptor family 52 subfamily N member 4 (gene/pseudogene) | OR52N4 | OR11-64 | 25.4kb | Anti-sense | Chr. 11 - 5722137 - 5755958 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
