Assay Details
Gene Symbol: GAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011519972.2 | XP_011518274.1 | 1 | 140 | 85 | |
GenBank mRNA | BC005077.1 | - | 1 | 749 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1495 | LINC01495 | -133.5kb | Anti-sense | Chr. 11 - 22445672 - 22492019 on NCBI Build GRCh38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011519972.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
