Assay Details
Gene Symbol: LOC100422737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ420564.1 | - | 1 | 346 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1 | QRSL1 | GatA | -56.9kb | Anti-sense | Chr. 6 - 106629566 - 106668417 on NCBI Build GRCh38 |
Downstream | microRNA 587 | MIR587 |
MIRN587, hsa-mir-587 |
57.0kb | Anti-sense | Chr. 6 - 106784125 - 106784220 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
