Assay Details
Gene Symbol: DCAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK026213.1 | - | 1 | 1050 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | phosphoprotein enriched in astrocytes 15 | PEA15 |
HMAT1, HUMMAT1H, MAT1, MAT1H, PEA-15, PED |
-46.4kb | Sense | Chr. 1 - 160205319 - 160215376 on NCBI Build GRCh38 |
Downstream | peroxisomal biogenesis factor 19 | PEX19 |
D1S2223E, HK33, PBD12A, PMP1, PMPI, PXF, PXMP1 |
13.3kb | Anti-sense | Chr. 1 - 160276809 - 160285151 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
