Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028000.hg.1 |
TC05002012.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028000.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028000.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028000.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027999.hg.1 |
TC05002012.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027999.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027999.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027999.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012667.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188457.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188457.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188457.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188457.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188457.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188457.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188457.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188457.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188457.hg.1 |
TC0500012667.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012667.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188456.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188456.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188456.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188456.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188456.hg.1 |
ENST00000518301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188456.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188456.hg.1 |
ENST00000518580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188456.hg.1 |
NR_027111.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188456.hg.1 |
TC0500012667.hg.1 |
Clariom D TC |