Assay Details
Gene Symbol: LOC285629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027111.1 | 2 - 3 | 293 | 61 | ||
GenBank mRNA | AK090625.1 | - | 2 - 3 | 293 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ATPase phospholipid transporting 10B (putative) | ATP10B | ATPVB | -2.4kb | Sense | Chr. 5 - 160563120 - 160929411 on NCBI Build GRCh38 |
Downstream | gamma-aminobutyric acid type A receptor beta2 subunit | GABRB2 | 352.3kb | Sense | Chr. 5 - 161288429 - 161548124 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05002012.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413416 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413451 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592929 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592937 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592959 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413464 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413423 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413375 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413420 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413388 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592944 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592923 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592936 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592914 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592947 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413376 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413394 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413438 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413446 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592986 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413377 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592918 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15757594 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592916 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15757599 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413407 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413432 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592968 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413397 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413427 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592992 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413393 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413448 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413455 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413408 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592928 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413441 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413444 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592990 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413418 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592932 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413386 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413392 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413398 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413440 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592975 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413410 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413439 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592988 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413413 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592919 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15757589 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413402 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592933 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413424 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15757592 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592965 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592964 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413400 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413426 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413452 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413461 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592955 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413385 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413463 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592967 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592969 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04413447 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24592943 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 837238 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 352716 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 64325 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5014 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5015 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413463 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592994 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413461 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592990 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592986 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592983 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592992 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413452 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592984 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413464 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413450 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592988 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413455 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592989 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592989 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592983 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592990 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413455 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413464 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413452 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592994 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592988 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592992 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592986 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413450 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413463 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04413461 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24592984 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P700572 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413451 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413386 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413434 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15757592 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413442 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413438 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413452 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413385 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413416 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413466 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413377 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413381 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413412 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413420 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413441 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413458 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413439 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15757596 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413440 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413399 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413401 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413411 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413463 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413465 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413432 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413407 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413408 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413405 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413379 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413414 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413392 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413417 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413444 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413410 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413424 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413423 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413426 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413383 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413457 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413403 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413429 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413447 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413396 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413461 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413389 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04413397 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15757594 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | ENST00000518301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | ENST00000518301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | ENST00000518301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012667.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05027997.hg.1 | ENST00000518301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05027997.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05027997.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05027997.hg.1 | TC05002012.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05027998.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05027998.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05027998.hg.1 | TC05002012.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | ENST00000518301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | ENST00000518301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | ENST00000518301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012667.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500188454.hg.1 | ENST00000518301.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500188454.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500188454.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500188454.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500188454.hg.1 | TC0500012667.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500188454.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500188454.hg.1 | ENST00000518301.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500188454.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500188454.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500188455.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500188455.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500188455.hg.1 | NR_027111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500188455.hg.1 | TC0500012667.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500188455.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500188455.hg.1 | ENST00000518580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500188455.hg.1 | NR_027111.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A027659 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A027659 | - | - |
