Assay Details
Gene Symbol: LINC00879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015400.1 | 2 - 3 | 259 | 86 | ||
GenBank mRNA | BC038721.1 | - | 2 - 3 | 262 | 86 | |
BC048119.1 | - | 2 - 3 | 260 | 86 | ||
BC115359.1 | - | 2 - 3 | 178 | 86 | ||
BC115360.1 | - | 2 - 3 | 211 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NOP2/Sun RNA methyltransferase family member 3 | NSUN3 |
MST077, MSTP077 |
-836.1kb | Sense | Chr. 3 - 94063011 - 94126786 on NCBI Build GRCh38 |
Downstream |
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase pseudogene 1 |
MTHFD2P1 | 683.1kb | Anti-sense | Chr. 3 - 95654423 - 95683193 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000476.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000476880.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | ENST00000476880.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03009167.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009167.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009167.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03009167.hg.1 | TC03000476.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03009168.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009168.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03009168.hg.1 | TC03000476.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000462219.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000470465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000476880.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | ENST00000476880.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008059.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157472.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157472.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157472.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157472.hg.1 | TC0300008059.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300157472.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157472.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157472.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157471.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157471.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157471.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157471.hg.1 | NR_015400.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157471.hg.1 | TC0300008059.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300157471.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157471.hg.1 | ENST00000463200.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157471.hg.1 | ENST00000466560.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157471.hg.1 | NR_015400.1 | Refseq |
