Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007793.hg.1 |
TC06000641.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007793.hg.1 |
ENST00000371197.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007793.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008211.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157435.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157435.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157435.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157435.hg.1 |
ENST00000371197.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157435.hg.1 |
ENST00000545705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157435.hg.1 |
NM_001010904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157435.hg.1 |
TC0600008211.hg.1 |
Clariom D TC |