Assay Details
Gene Symbol: LOC101928565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF087964.1 | - | 1 | 246 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100505918 | LOC100505918 | -62.4kb | Sense | Chr. 1 - 168400188 - 168422656 on NCBI Build GRCh38 | |
Downstream | X-C motif chemokine ligand 2 | XCL2 |
SCM-1b, SCM1B, SCYC2 |
55.4kb | Sense | Chr. 1 - 168540765 - 168543997 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
