Assay Details
Gene Symbol: ZAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017004867.1 | XP_016860356.1 | 1 | 82 | ||
XM_017004868.1 | XP_016860357.1 | 1 | 82 | |||
XM_017004869.1 | XP_016860358.1 | 1 | 82 | |||
XM_017004870.1 | XP_016860359.1 | 1 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1125 | LINC01125 | -9.7kb | Anti-sense | Chr. 2 - 97669743 - 97703066 on NCBI Build GRCh38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017004867.1, XM_017004868.1, XM_017004869.1, XM_017004870.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
