Assay Details
Gene Symbol: MAP3K13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC026249.1 | - | 1 | 2449 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase |
EHHADH |
ECHD, FRTS3, L-PBE, LBFP, LBP, PBFE |
-80.2kb | Anti-sense | Chr. 3 - 185190624 - 185282886 on NCBI Build GRCh38 |
Downstream | transmembrane protein 41A | TMEM41A | 2900010K02Rik | 117.0kb | Anti-sense | Chr. 3 - 185482671 - 185499057 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
