Assay Details
Gene Symbol: LOC101928730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120422.1 | 1 - 2 | 746 | 112 | ||
GenBank mRNA | BC034786.1 | - | 1 - 2 | 746 | 112 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1070 | LINC01070 | -70.5kb | Anti-sense | Chr. 13 - 112197333 - 112201000 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 1043 | LINC01043 | 40.1kb | Anti-sense | Chr. 13 - 112313467 - 112321766 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
