Assay Details
Gene Symbol: HNF4A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC071794.1 | - | 1 - 2 | 364 | 83 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BG427859.1 | - | 366 | Chr. - - [-] on Build | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | R3H domain containing like | R3HDML | dJ881L22.3 | -40.6kb | Anti-sense | Chr. 20 - 44337158 - 44350792 on NCBI Build GRCh38 |
Downstream | microRNA 3646 | MIR3646 | 5.3kb | Anti-sense | Chr. 20 - 44408120 - 44408203 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
