Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009118.hg.1 |
ENST00000556913.1 |
Ensembl |