Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154264.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154264.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154264.hg.1 |
TC1300008067.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300154264.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154264.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154267.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154267.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154267.hg.1 |
TC1300008067.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300154267.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154267.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008067.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154265.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154265.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154265.hg.1 |
TC1300008067.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300154265.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154265.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154272.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154272.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154272.hg.1 |
TC1300008067.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300154272.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154272.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154271.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154271.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154271.hg.1 |
TC1300008067.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300154271.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154271.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154269.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154269.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154269.hg.1 |
TC1300008067.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300154269.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154269.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154270.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154270.hg.1 |
NR_108094.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154270.hg.1 |
TC1300008067.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300154270.hg.1 |
ENST00000561786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154270.hg.1 |
NR_108094.1 |
Refseq |