Assay Details
Gene Symbol: LOC101927078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_130785.1 | 11 - 12 | 1546 | 84 | ||
GenBank mRNA | AK097686.1 | - | 11 - 12 | 1541 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | YTH domain containing 2 | YTHDC2 | CAHL | -853.3kb | Anti-sense | Chr. 5 - 113513683 - 113595287 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101927059 | LOC101927059 | 106.2kb | Anti-sense | Chr. 5 - 114576041 - 114579970 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
