Assay Details
Gene Symbol: LOC100506016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134944.1 | 1 - 2 | 1081 | 78 | ||
GenBank mRNA | BC038751.1 | - | 1 - 2 | 1082 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 403 | LINC00403 | -47.1kb | Anti-sense | Chr. 13 - 111972310 - 112108015 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 1070 | LINC01070 | 36.6kb | Anti-sense | Chr. 13 - 112197333 - 112201000 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
