Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400161560.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161560.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400161560.hg.1 |
TC0400008962.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400161560.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400161560.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161561.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161561.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400161561.hg.1 |
TC0400008962.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400161561.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161561.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
ENST00000503100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008962.hg.1 |
NR_125887.1 |
Refseq |