Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011241.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011241.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011241.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011241.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011241.hg.1 |
NR_110809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011241.hg.1 |
NR_110809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011241.hg.1 |
NR_110809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011241.hg.1 |
NR_110809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011241.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011241.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011241.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011241.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011241.hg.1 |
NR_110809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011241.hg.1 |
NR_110809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011241.hg.1 |
NR_110809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011241.hg.1 |
NR_110809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190334.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190334.hg.1 |
NR_110809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190334.hg.1 |
TC1700011241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700190334.hg.1 |
NR_110809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190334.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190335.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190335.hg.1 |
NR_110809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190335.hg.1 |
TC1700011241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700190335.hg.1 |
ENST00000584805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190335.hg.1 |
NR_110809.1 |
Refseq |