Assay Details
Gene Symbol: LOC105375532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX648938.1 | - | 1 | 7350 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | homeodomain interacting protein kinase 2 | HIPK2 | PRO0593 | -161.8kb | Anti-sense | Chr. 7 - 139561570 - 139777894 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC105375531 | LOC105375531 | 23.2kb | Sense | Chr. 7 - 139970393 - 139978524 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
