Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200169741.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200169741.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200169741.hg.1 |
TC1200009358.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200169741.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200169741.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200169744.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200169744.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200169744.hg.1 |
TC1200009358.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200169744.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200169744.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200169739.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200169739.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200169739.hg.1 |
TC1200009358.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200169739.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200169739.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
ENST00000541258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009358.hg.1 |
NR_104648.1 |
Refseq |