Assay Details
Gene Symbol: LOC105376037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK126527.1 | - | 1 | 3549 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | exosome component 3 | EXOSC3 |
CGI-102, PCH1B, RRP40, Rrp40p, bA3J10.7, hRrp-40, p10 |
-3.9kb | Sense | Chr. 9 - 37779714 - 37785092 on NCBI Build GRCh38 |
Downstream | DDB1 and CUL4 associated factor 10 | DCAF10 | WDR32 | 7.9kb | Anti-sense | Chr. 9 - 37800554 - 37867669 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
