Assay Details
Gene Symbol: PDZRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK130896.1 | - | 1 | 1045 | 109 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein phosphatase 4 regulatory subunit 2 | PPP4R2 | PP4R2 | -446.7kb | Anti-sense | Chr. 3 - 72996743 - 73069201 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101927296 | LOC101927296 | 7.1kb | Anti-sense | Chr. 3 - 73521322 - 73569774 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
