Assay Details
Gene Symbol: WWTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK127620.1 | - | 1 | 2145 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane 4 L six family member 4 | TM4SF4 |
ILTMP, il-TMP |
-67.8kb | Anti-sense | Chr. 3 - 149474581 - 149503394 on NCBI Build GRCh38 |
Downstream | WWTR1 antisense RNA 1 | WWTR1-AS1 | 84.6kb | Anti-sense | Chr. 3 - 149658210 - 149661364 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
