Assay Details
Gene Symbol: ANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK128049.1 | - | 1 | 348 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ribosomal protein S26 pseudogene 25 | RPS26P25 | RPS26_10_512 | -155.4kb | Anti-sense | Chr. 4 - 113213969 - 113214420 on NCBI Build GRCh38 |
Downstream | calcium/calmodulin dependent protein kinase II delta | CAMK2D | CAMKD | 80.3kb | Anti-sense | Chr. 4 - 113451032 - 113761927 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
