Assay Details
Gene Symbol: LOC105373460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL117577.1 | - | 1 | 1054 | 96 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | DA572782.1 | - | 1 | Chr. 2 - 19488684 - 19489466 [-] on Build GRCh38 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928196 | LOC101928196 | -19.3kb | Anti-sense | Chr. 2 - 19398651 - 19468822 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 954 | LINC00954 | 379.5kb | Sense | Chr. 2 - 19868854 - 19885047 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
