Assay Details
Gene Symbol: LOC105370474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC038722.1 | - | 1 | 253 | 111 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 179 member B | FAM179B | KIAA0423 | -9.0kb | Anti-sense | Chr. 14 - 44962190 - 45074431 on NCBI Build GRCh38 |
Downstream | pre-mRNA processing factor 39 | PRPF39 | 122.0 | Anti-sense | Chr. 14 - 45084099 - 45115601 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
