Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006798.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006798.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006798.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006798.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006798.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006798.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006798.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006798.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006798.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006798.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300147502.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300147502.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300147502.hg.1 |
TC0300006798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300147502.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300147502.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300147498.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300147498.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300147498.hg.1 |
TC0300006798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300147498.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300147498.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300147496.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300147496.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300147496.hg.1 |
TC0300006798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300147496.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300147496.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300147493.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300147493.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300147493.hg.1 |
TC0300006798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300147493.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300147493.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006798.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006798.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006798.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006798.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006798.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006798.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006798.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006798.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006798.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006798.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300147501.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300147501.hg.1 |
NR_110814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300147501.hg.1 |
TC0300006798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300147501.hg.1 |
ENST00000455626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300147501.hg.1 |
NR_110814.1 |
Refseq |