Assay Details
Gene Symbol: LOC102723648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC042825.1 | - | 4 | 449 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mitogen-activated protein kinase 13 | MAPK13 |
MAPK 13, MAPK-13, PRKM13, SAPK4, p38delta |
-13.5kb | Anti-sense | Chr. 6 - 36130484 - 36144524 on NCBI Build GRCh38 |
Downstream | bromodomain and PHD finger containing 3 | BRPF3 | 37.8kb | Anti-sense | Chr. 6 - 36196773 - 36232790 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
